Would you like to log in again?
Rat Recombinant IL-1 Beta
Supplier: Rockland Immunochemicals
012-001-301-0100 012-001-301-1000 012-001-301-0010 012-001-301-0002
75929-000EA 326 USD InStock
75929-000 75929-002 75929-004 75928-998
Rat Recombinant IL-1 Beta
Proteins and Peptides
Recombinant Rat IL-1 beta control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-1 beta (IL-1β) is a pro-inflammatory cytokine, produced in response to inflammatory agents by a variety of cells, including, monocytes, macrophages, and dendritic cells (DCs). IL-1β and IL-1α are two distinct and independently regulated gene products that comprise IL-1 and signal through the Type 1 IL-1 receptor (IL-1R1). Although IL-1α is cell associated and IL-1β is secreted, they have nearly identical biological activity in that they induce adhesion molecule expression on epithelial cells, control fever induction, and play a role in arthritis and septic shock. Signaling activated by the IL-1R1 promotes these activities through a MYD88 signaling pathway similar to those associated with Toll receptors. Recombinant rat IL-1β is a non-glycosylated protein, containing 152 amino acids, with a molecular weight of 17.3 kDa.
Interleukin-1-beta Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-1-beta in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty