Would you like to log in again?
Rat Recombinant FGF-9
Supplier: Rockland Immunochemicals
012-001-U76-1000 012-001-U76-0002 012-001-U76-0010 012-001-U76-0100
10801-382EA 6098 USD InStock
10801-382 10802-876 10802-872 10802-874
Rat Recombinant FGF-9
Proteins and Peptides
Recombinant Rat FGF-9 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Fibroblast Growth Factor-9 (FGF-9) is a steroid-regulated mitogen and survival factor for nerve and mesenchymal cells. FGF-9 is an autocrine/paracrine growth factor considered to be important for the growth and survival of motorneurons and prostate tissue. Recombinant rat FGF-9 is a non-glycosylated protein, containing 207 amino acids, with a molecular weight of 23.3 kDa.
Fibroblast Growth Factor 9 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Fibroblast Growth Factor 9 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty