Would you like to log in again?
Rat Recombinant CCL2
Supplier: Rockland Immunochemicals
012-001-V74-0010 012-001-V74-0100 012-001-V74-1000 012-001-V74-0002
75930-026EA 326 USD InStock
75930-026 75930-034 75930-022 75930-030
Rat Recombinant CCL2
Proteins and Peptides
Recombinant Rat CCL2 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Monocyte Chemotactic Protein 1 (MCP-1), also known as CCL2, is thought to be produced by injured or infected tissues. MCP-1 signals through G protein-coupled receptors, CCR2 and CCR4, to recruit memory T cells, monocytes and dendritic cells. Recombinant rat MCP-1is a non-glycosylated protein, containing 125 amino acids, with a molecular weight of 14.1 kDa.
Monocyte Chemotactic Protein-1 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Monocyte Chemotactic Protein-1 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty