Would you like to log in again?
Rat SCF
Supplier: Rockland Immunochemicals
012-001-W20-0100 012-001-W20-1000 012-001-W20-0002 012-001-W20-0010
75932-256EA 4582 USD InStock
75932-256 75932-268 75932-262 75932-274
Rat SCF
Proteins and Peptides
Recombinant Rat MIP-1 alpha (CCL3) control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Macrophage Inflammatory Protein-1 alpha (MIP-1α), also known as CCL3, is produced by macrophages and is thought to induce inflammatory responses, including superoxide production by neutrophils. MIP-1α can exist as a naturally occurring heterodimer with MIP-1β and has been shown to have antiviral activity against HSV-1. Recombinant rat MIP-1α is a non-glycosylated protein, containing 69 amino acids, with a molecular weight of 7.8 kDa.
Macrophage Inflammatory Protein-1 alpha Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Macrophage Inflammatory Protein-1 alpha in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty