Would you like to log in again?
Rat Tf
Supplier: Rockland Immunochemicals
012-0134
75935-202EA 265 USD InStock
75935-202
Rat Tf
Proteins and Peptides
Rat Transferrin
  • control or standard reagent
  • SDS, Western Blotting, and ELISA experiments
Rat Transferrin is a blood plasma glycoprotein which binds iron and regulates the amount of available iron in circulation. Rat Transferrin has a high affinity for iron, but this binding is reversible. Rat Transferrin has a molecular weight of approximately 80kDa. Rat Transferrin also plays a role in the immune system, creating environments low in iron for which many pathogenic bacteria are unable to thrive.
Rat Transferrin is a suitable protein for use as a control reagent in both Western Blotting and ELISA experiments. For blotting, the expected molecular weight of Rat Transferrin is approximately 80kDa.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty