Would you like to log in again?
Rat Recombinant FGF-Basic
Supplier: Rockland Immunochemicals
012-F01-U80-1000 012-F01-U80-0010 012-F01-U80-0050
10802-880EA 148 USD InStock
10802-880 10802-878 10802-882
Rat Recombinant FGF-Basic
Proteins and Peptides
Recombinant Rat FGF-basic control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Fibroblast Growth Factors (FGFs) are a 22 member family of proteins known to be involved in angiogenesis, wound healing and embryonic development. As a family, they bind to heparin and signal through four receptor tyrosine kinases called, FGFR1, 2, 3 and 4. Recombinant rat FGF-basic is a non-glycosylated protein, containing 146 amino acids, with a molecular weight of 16.4 kDa.
Fibroblast Growth Factor basic Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Fibroblast Growth Factor basic in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty