Would you like to log in again?
Sheep IgG (AP (Alkaline Phosphatase))
Supplier: Rockland Immunochemicals
013-0502
RL013-0502EA 108 USD InStock
RL013-0502
Sheep IgG (AP (Alkaline Phosphatase))
Antibodies
Sheep IgG Alkaline Phosphatase Conjugate
  • control or standard reagent
  • ELISA, western blotting, Immunohistochemistry
Sheep IgG Alkaline Phosphatase Conjugated Whole Molecule - secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. The whole IgG molecule possesses both the F(c) region, recognized by high-affinity Fc receptor proteins, as well as the F(ab) region possessing the epitope-recognition site. Both heavy and light chains of the antibody molecule are present.
Sheep IgG Alkaline Phosphatase Conjugated Whole Molecule Secondary Antibody reagents are ideal for ELISA, western blotting, Immunohistochemistry, as well as other antibody detection methods.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty