Would you like to log in again?
Armenian Hamster IgG Isotype Control
Supplier: Rockland Immunochemicals
019-001-002
RL019-001-002EA 196.04 USD InStock
RL019-001-002
Armenian Hamster IgG Isotype Control
Antibodies
Armenian Hamster IgG Whole Molecule isotype Control
  • control or standard reagent
  • SDS, Western Blotting, and ELISA experiments
Armenian Hamster isotype controls are used in flow cytometry, western blot and ELISA and differentiate between immunoglobulin classes and subclasses. Isotype controls allow for the genetic variations or differences in the constant regions of the heavy and light chains. In hamster there are six relevant heavy chain isotypes and two light chain isotypes: heavy chain alpha - IgA, gamma - IgG 1, 2a, 2b, 3 and μ - IgM, light chain kappa and lambda.
ARMENIAN HAMSTER IgG whole molecule can be utilized as a control or standard reagent in Western Blotting and ELISA experiments. Specific conditions should be optimized by user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty