Would you like to log in again?
Bovine (Thyroidectomized) Serum Sterile, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
D118-00-0100 D118-00-0050
10802-170EA 621 USD InStock
10802-170 10802-168
Bovine (Thyroidectomized) Serum Sterile, Rockland Immunochemicals
Sera (Serum)
Thyroidectomized Sterile Serum
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Bovine Serum / Calf Serum is used as a supplement to cell culture media. Bovine Serum is also suitable for use as a component of bioassays, immunoassays or enzyme assays. Bovine Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation. Bovine Serum is ideal for investigators in Cancer, Immunology, and Cell Biology research.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty