Would you like to log in again?
Mouse (Virus Free) Serum Sterile, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
D123-00
75930-114EA 886.8 USD InStock
75930-114
Mouse (Virus Free) Serum Sterile, Rockland Immunochemicals
Sera (Serum)
Sterile Serum
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Mouse Serum is used as a supplement to cell culture media. Mouse Serum is also suitable for use as a component of bioassays, immunoassays or enzyme assays. Mouse Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation. Mouse Serum is ideal for investigators in Cancer, Immunology, and Cell Biology research.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty