Would you like to log in again?
Rabbit Serum Non sterile, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
D209-00-0050 D209-00-0100
75934-130EA 534 USD InStock
75934-130 75934-128
Rabbit Serum Non sterile, Rockland Immunochemicals
Sera (Serum)
Non-Sterile Serum
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Rabbit Serum is used as a component of bioassays, immunoassays or enzyme assays. Rabbit Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Rabbit Serum is also available as a sterile preparation for use as a cell culture supplement. Rabbit Serum is ideal for investigators in Cancer, Immunology and Cell Biology research. Rabbit Serum can be used as a blocking agent to treat plastic surfaces, membrane or tissue after they have been sensitized with primary antibody or antigen. It provides an alternative to fish gel and other blocking agents. It is effective in reducing nonspecific binding of proteins to reaction surfaces, thereby maximizing signal-to-noise ratio. This blocking agent is recommended for use in immunoassays where the primary antibody was produced in mouse, as a source of non-specific serum protein or on tissue for immunohistochemical applications.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty