Would you like to log in again?
Turkey Serum Non-Sterile, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
D213-00
75933-282EA 487 USD InStock
75933-282
Turkey Serum Non-Sterile, Rockland Immunochemicals
Sera (Serum)
Non-Sterile Serum
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Turkey Serum is used as a component of bioassays, immunoassays or enzyme assays. Turkey Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Turkey Serum is also available as a sterile preparation for use as a cell culture supplement.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty