Would you like to log in again?
Chicken Serum (Lyophilized), Rockland Immunochemicals
Supplier: Rockland Immunochemicals
D302-05
10802-362EA 140.93 USD InStock
10802-362
Chicken Serum (Lyophilized), Rockland Immunochemicals
Sera (Serum)
Lyophilized Non-Sterile Serum
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Chicken Serum is used as a supplement to cell culture media. Chicken Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Chicken Serum is ideal for investigators in Cancer and Cell Biology.
Suitable for use as a control or diluent in assays and most other immunological methods.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty