Would you like to log in again?
Mouse C57 Black Plasma Non Sterile In Sodium Citrate, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
D424-03
75930-132EA 4344.98 USD InStock
75930-132
Mouse C57 Black Plasma Non Sterile In Sodium Citrate, Rockland Immunochemicals
Blood
Non-Sterile Plasma in Anticoagulant
  • prepared by collection of whole blood into said anticoagulant
  • Red blood cells are removed through a process of centrifugation and filtration techniques - leaving non-sterile plasma behind
used as a component of bioassays, immunoassays or enzyme assays
provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health
also available as a sterile preparation for use as a cell culture supplement
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty