Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Duck Plasma Sterile In Potassium EDTA, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
D514-05
10801-014EA 573.79 USD InStock
10801-014
Duck Plasma Sterile In Potassium EDTA, Rockland Immunochemicals
Blood
Sterile Plasma in Anticoagulant
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by collection of whole blood into said anticoagulant
  • Red blood cells are removed through a process of centrifugation and filtration techniques - leaving non-sterile plasma behind
used as a component of bioassays, immunoassays or enzyme assays
used as a supplement to cell culture media
provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health
Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty