Would you like to log in again?
GFP Western Blot Kit: for GFP Chemiluminescent Western Blotting, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KCA215
RLKCA215EA 382 USD InStock
RLKCA215
GFP Western Blot Kit: for GFP Chemiluminescent Western Blotting, Rockland Immunochemicals
Assays Immunological Assays
Green Fluorescent Protein (GFP) Chemiluminescent Western Blot Kit
  • Chemiluminescent Western Blot Kit for GFP contains all of the necessary reagents
  • rapid proven protocol - includes straightforward procedures
  • extremely high signal detection of our luminol chemiluminescent substrate for the detection of recombinant proteins containing GFP and its variants
  • This kit contains sufficient substrate for up to 30 mini blots at 7.5 x 8 cm2 (1,800 cm2) and is stable for at least 1 year when stored as indicated.
GFP Chemiluminescent Western Blot Kit allows for the detection of GFP-tagged recombinant proteins present in cell lysates provided by the user. After protein separation by SDS-PAGE and transfer, the membrane is probed with monoclonal Anti-GFP. Detection of the membrane bound antibody-antigen complex is achieved by the addition of a secondary antibody conjugated to the enzyme horseradish peroxidase. The enzyme reacts with a specialized formulation of luminol, an extremely sensitive, non-radioactive substrate that emits light and allows visualization using X-ray film or other imaging methods, including highly sensitive CCD cameras and imaging systems.
GFP Western Blot Kit is ideal for detection of GFP-tagged recombinant proteins by western blot. The GFP Western Blot kit makes use of Rockland's optimized anti-GFP antibody. The Chemiluminescent GFP Western Blot kit is useful for both western blotting and dot blotting methods. Please read the entire product insert prior to use.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty