Would you like to log in again?
Isotyping Kit: Rapid Mouse Monoclonal Isotyping Kit IsoMax™, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KDA-005 KDA-010
75934-792EA 292 USD InStock
75934-792 75934-794
Isotyping Kit: Rapid Mouse Monoclonal Isotyping Kit IsoMax™, Rockland Immunochemicals
Assays Immunological Assays
Rapid Mouse Monoclonal Isotyping Kit IsoMax™
  • contains 5 or 10 packs of 2 cassettes each:
  • one cassette for detecting IgG1, IgG2a and IgG2b; the other cassette for detecting IgG3, IgA and IgM
  • a rapid (5 minute) lateral flow assay with sensitivity equal to ELISA for monoclonal antibody class and subclass determination
  • test antibody present in tissue culture supernatant or mouse ascites fluid
Use Rockland Immunochemicals' IsoMax™ Rapid Mouse Monoclonal Isotyping Kit for determining the class and subclass of a monoclonal antibody. Isotype determination is a prerequisite for selecting the best immunoglobulin purification method. For example, IgA and IgM are often best purified by size (gel filtration) or on immunoaffinity support columns, whereas IgG2a and IgG2b can be purified by protein A chromatography at a pH of 7 to 8. IgG1 binds best to protein A at a pH of 8 to 9. Isotype determination is also required if an antibody is to be digested by pepsin or papain as each isotype shows varying sensitivity to enzymatic digestion. Please call technical service for further assistance.
Each kit contains 5 or 10 packs of 2 cassettes each: one cassette for detecting IgG1, IgG2a and IgG2b; the other cassette for detecting IgG3, IgA and IgM. When a properly diluted sample containing a specific isotype antibody is added to the sample well, a specific class and subclass soluble complex is formed with the embedded gold conjugates. These complexes travel the length of the membrane and are resolved on the anti-isotype and anti-class-specific antibody impregnated membrane. A control line will appear on the membrane in the region marked ''C'' on the cassette indicating a successful run (positive internal control). Typically when antibodies are applied at a concentration of 10 ng/mL the best results are obtained by reading the cassette 5 to 10 minutes after application. Results should NOT be read after 10 minutes.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty