Would you like to log in again?
RadChemDosi™ Cell Survival Assay Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KLD-003
75931-428EA 471 USD InStock
75931-428
RadChemDosi™ Cell Survival Assay Kit, Rockland Immunochemicals
Assays Cellular Assays
RadChemDosi™ Cell Survival Assay Kit
  • RadChemDosi™ Cell Survival Assay Kit contains enough reagents for 100 test if 1 ml is used for irradiation.
RadChemDosi™ Cell Survival Assay Kit is a chemical based assay that can measure the dose rate of ionizing radiation. Radiation mainly works by producing free radicals that reacts with biomolecules (DNA, protein and lipid) in mammalian and other types of cells. The amount of free radicals produced by radiation is dependent on the dose. RadChemDosi™ is an effective way to measure dose rate for ionizing radiation. It is a chemical-based assay that uses the ability of ionizing radiation to induce changes in certain chemicals. RadChemDosi™ is useful for blood irradiators where it can be used for periodic calibration and to determine the dose given to blood by simultaneously irradiating RadChemDosi™ in small disposable tubes attached to the blood sample by taping the tubes outside the bag.
RadChemDosi™ Cell Survival Assay Kit contains enough reagents for 100 test if 1 ml is used for irradiation. RadChemDosi™ is sensitive to measure the dose rate as low as 2.5Gy/min at various positions in a Cesium irradiator chamber. In addition to calibrating the dose rate at each position in a radiation chamber periodically, it can also be used to measure the dose rate in each well of a 6 and 24 wells plate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty