Would you like to log in again?
Human ANG ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0145
76236-594EA 687 USD InStock
76236-594
Human ANG ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human ANG AccuSignal ELISA Kit
  • Natural and recombinant human ANG.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: Q25-P147
Angiogenin(Ang) also known as ribonuclease 5 is a protein that in humans is encoded by the ANG gene. Angiogenin is a potent stimulator of new blood vessel formation. It hydrolyzes cellular tRNAs resulting in decreased protein synthesis and is similar to pancreatic ribonuclease. Hooper et al.(2003) reviewed the evidence that angiogenins are involved in host defense and noted that inflammation provokes upregulated ANG mRNA expression in liver and an increase in detectable ANG protein in serum. Weremowicz et al.(1989, 1990) assigned the human angiogenin gene to chromosome 14q11.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml, 78pg/ml human ANG standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human ANG standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty