Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Mouse EPO ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0162
76237-136EA 723.6 USD InStock
76237-136
Mouse EPO ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse EPO AccuSignal ELISA Kit
  • Natural and recombinant mouse EPO.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A27-R192
Erythropoietin, EPO, also known as hematopoietin or hemopoietin, is a glycoprotein hormone that controls erythropoiesis, or red blood cell production. It is a cytokine for erythrocyte(red blood cell) precursors in the bone marrow. Its gene is mapped to 7q22. It is said that the EPO gene encodes a deduced 193-amino acid propolypeptide. This hormone can be found in kidney and liver. It is the hormone that regulates red blood cell production. And it plays an important role in the brain's response to neuronal injury. What's more, EPO is also involved in the wound healing process.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 3000pg/ml, 1500pg/ml, 750pg/ml,,419.pg/ml, 187.5pg/ml, 93.8pg/ml, 46.9pg/ml mouse EPO standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernatants, serum, plasma(heparin) or tissue lysates to each empty well. We recommend that each mouse EPO standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty