Would you like to log in again?
Human IGFBP4 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0205
76236-634EA 636 USD InStock
76236-634
Human IGFBP4 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human IGFBP4 AccuSignal ELISA Kit
  • Natural and recombinant human IGFBP4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: D22-E258
Insulin-like growth factor-binding protein 4 is a protein that in humans is encoded by the IGFBP4gene. This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma in both glycosylated and non-glycosylated forms. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors. In addition, IGFBP-4 is a unique protein and it consistently inhibits several cancer cells in vivo and in vitro. Its inhibitory action has been shown in vivo in prostate and colon. It is secreted by all colon cancer cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 32000pg/ml, 16000pg/ml, 8000pg/ml, 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml human IGFBP4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human IGFBP4 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty