Would you like to log in again?
Human IL-3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0218
76236-660EA 636 USD InStock
76236-660
Human IL-3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-3 AccuSignal ELISA Kit
  • Natural and recombinant human IL-3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A20-F152
The gene IL-3 encodes interleukin 3, a hematopoietic colony-stimulating factor(CSF) that is capable of supporting the proliferation of a broad range of hematopoietic cell types. Interleukin-3(IL-3), a protein of 140 amino acids, is chemically synthesized by means of an automated peptide synthesizer and is shown to have the biological activities attributed to native IL-3. The cDNA sequence for murine interleukin-3, one of the colony stimulating factors that regulate haematopoiesis, codes for a polypeptide of 166 amino acids including a putative signal peptide. The mouse IL 3 gene is located on chromosome 11. The human gene encoding IL 3 is tandemly arrayed on the long arm of chromosome 5. The standard product used in this kit is recombinant human IL-3, consisting of 133 amino acids with the molecular mass of 15KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human IL-3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma( heparin, EDTA, citrate) to each empty well. It is recommended that each human IL-3 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty