Would you like to log in again?
Human IL-15 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0239
76236-694EA 642 USD InStock
76236-694
Human IL-15 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-15 AccuSignal ELISA Kit
  • Natural and recombinant human IL-15.
  • There is no detectable cross-reactivity with other relevant proteins
  • Expression system for standard: E.coli; Immunogen sequence: N49-S162
Interleukin(IL)-15 is a cytokine with the ability to stimulate the proliferation activity of Th1 and/or Th2 lymphocytes. IL-15 is a novel cytokine whose effects on T-cell activation and proliferation are similar to those of interleukin-2(IL-2), presumably because IL-15 utilizes the beta and gamma chains of the IL-2 receptor. IL-15 can play a role in the initiation and outcome of acute and chronic rejection. Anti-IL-15 therapy in combination with classic immunosuppression therapy might thus be beneficial in the prevention of acute, and especially chronic, allograft rejection. The human IL15 gene is mapped to human chromosome 4q31 by fluorescence in situ hybridization.1 The standard product used in this kit is recombinant human IL-15, consisting of 114 amino acids with the molecular mass of 12.9KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml human IL-15 standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human IL-15 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty