Would you like to log in again?
Rat Laminin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0244
76236-704EA 839.55 USD InStock
76236-704
Rat Laminin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat Laminin AccuSignal ELISA Kit
  • Natural rat Laminin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • From rat sarcoma basement membrane
Laminin is a large basement membrane glycoprotein composed of three subunits designated the A, B1, and B2. Laminin has diverse biological functions, which include stimulating epithelial cell growth and differentiation. The nucleotide sequence of human laminin A chain has an open reading frame encoding 3075-amino acids. The human laminin A chain is at locus 18p11.3. The nucleotide sequence of the human laminin B1 reveals a 5358-base pair open reading frame that potentially codes for 1786 amino acids, including 20 amino acids of a presumptive signal peptide. The gene for the human laminin-B1 chain has been localized to chromosome 7, band q31. The B2 chain consists of six distinct domains, including two domains with alpha-helical, coiled-coil structures, two domains with cysteine-rich homologous repeats, and two globular domains. The amino acid sequences of the B2 and B1 chains demonstrate considerable homology. The human laminin B2 chain gene maps to the long arm of chromosome 1 in the band q31.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml rat Laminin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each rat Laminin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty