Would you like to log in again?
Human CCL8 - MCP-2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0251
76236-714EA 636 USD InStock
76236-714
Human CCL8 - MCP-2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CCL8 - MCP-2 AccuSignal ELISA Kit
  • Natural and recombinant human CCL8.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence:  Q24-P99
Chemokine(C-C motif) ligand 8, also known as monocyte chemoattractant protein 2(MCP-2), is a protein that in humans is encoded by the CCL8 gene. CCL8 is a small cytokine belonging to the CC chemokine family. The CCL8 protein is produced as a precursor containing 109 amino acids, which is cleaved to produce mature CCL8 containing 75 amino acids. The gene for CCL8 is encoded by 3 exons and is located within a large cluster of CC chemokines on chromosome 17q11.2 in humans. Monocyte chemotactic protein 2(MCP-2) is a CC chemokine that utilizes multiple cellular receptors to attract and activate human leukocytes.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human CCL8 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CCL8 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty