Would you like to log in again?
Mouse CCL19 - MIP-3 beta ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0265
76236-734EA 635.74 USD InStock
76236-734
Mouse CCL19 - MIP-3 beta ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CCL19 - MIP-3 beta AccuSignal ELISA Kit
  • Natural and recombinant mouse CCL19.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence:  G26-S108
Chemokine(C-C motif) ligand 19 (CCL19) is a small cytokine belonging to the CC chemokine family that is also known as EBI1 ligand chemokine (ELC) and macrophage inflammatory protein-3-beta (MIP-3-beta). Using PCR of human-rodent hybrids, the CCL19 gene was mapped to human chromosome 9p13, and the CCL19 gene is not part of the CC chemokine gene cluster on chromosome 17. The cytokine encoded by this gene may play a role in normal lymphocyte recirculation and homing. It also plays an important role in trafficking of T cells in thymus, and in T cell and B cell migration to secondary lymphoid organs.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml mouse CCL19 standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse CCL19 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty