Would you like to log in again?
Human MMP-1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0266
76236-736EA 652.22 USD InStock
76236-736
Human MMP-1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human MMP-1 AccuSignal ELISA Kit
  • Natural and recombinant human total MMP-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence:  F20-N469
Matrix metalloproteinase-1(MMP-1) is also known as collagenase. Matrix metalloproteinases(MMPs) are zinc-dependent proteases that degrade extrMMP-1llular matrix proteins. The MMPs comprise a family of at least 20 proteolytic enzymes that play an essential role in tissue remodeling. Matrix metalloproteinase-1(MMP-1) plays an important role in the degradation of collagen in inflammatory diseases. MMP1(interstitial collagenase), MMP9(gelatinase B) and MMP12(macrophage elastase) are thought to be important in the development of emphysema. Smoking-induced MMP-1 might be important in the skin-ageing effects of tobacco smoking. The standard product used in this kit is human MMP-1 with the molecular mass of 51-53KDa. The detected MMP-1 includes zymogen and active enzyme.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human MMP-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma (heparin) to each empty well. It is recommended that each human MMP-1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty