Would you like to log in again?
Human MMP13 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0275
76236-754EA 686.86 USD InStock
76236-754
Human MMP13 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human MMP13 AccuSignal ELISA Kit
  • Natural and recombinant human total MMP13.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence:   L20-C471
Collagenase 3 is an enzyme that in humans is encoded by the MMP13 gene. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The protein encoded by this gene cleaves type II collagen more efficiently than types I and III. The expression of MMP13 in osteoarthritic cartilage and its activity against type II collagen indicates that the enzyme plays a significant role in cartilage collagen degradation and must, therefore, form part of a complex target for proposed therapeutic interventions based on collagenase inhibition. It may be involved in articular cartilage turnover and cartilage pathophysiology associated with osteoarthritis. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human MMP13 standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma( heparin) to each empty well. It is recommended that each human MMP13 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty