Would you like to log in again?
Human NGF - NGF beta ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0276
76236-756EA 635.74 USD InStock
76236-756
Human NGF - NGF beta ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human NGF - NGF beta AccuSignal ELISA Kit
  • Natural and recombinant human NGF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence:   S122-A241
Nerve growth factor(NGF) is a polypeptide involved in the regulation of growth and differentiation of sympathetic and certain sensory neurons. NGF is thought to have a profound effect on the development and maintenance of sympathetic and embryonic sensory neurones. NGF activity isolated from the male mouse submaxillary gland(MSG) consists of three types of subunits, alpha, beta and gamma, which specifically interact to form a 7S, approximately 130,000-molecular weight(Mr) complex. The 7S complex contains two identical 118-amino acid beta-chains, which are solely responsible for the nerve growth-stimulating activity of NGF. NGF, which is expressed by inflammatory cells and effects changes that lead to increased neural responsiveness, could be a pivotal mediator in allergic rhinitis. The standard product used in this kit is human 2.5S NGF, which is a dimmer linking with two polypeptide chains of 120 amino acids.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human NGF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernatants or serum to each empty well. It is recommended that each human NGF standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty