Would you like to log in again?
Mouse Neurotrophin-3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0280
76237-588EA 635.74 USD InStock
76237-588
Mouse Neurotrophin-3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Neurotrophin-3 AccuSignal ELISA Kit
  • Natural and recombinant mouse Neurotrophin-3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: Y139-T257
Neurotrophin-3(NT-3) is a new member of the nerve growth factor gene family, which plays an important role in the development and maintenance of the vertebrate nervous system.NT-3 and its receptor, called neurotrophic tyrosine kinase receptor type 3(Ntrk3; also called TrkC), are expressed early and throughout embryogenesis. NT-3 is one of five neurotrophin growth factors which shape the development of the nervous system by regulating neuronal survival and differentiation. NT-3 may be one of the central nervous system-derived factors that mediate neural crest(NC) cell proliferation in vivo. NT-3 has been mapped to human chromosome 12p and mouse chromosome 6.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse Neurotrophin-3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernatants or serum to each empty well. It is recommended that each mouse Neurotrophin-3 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty