Would you like to log in again?
Rat SCF ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0304
76237-186EA 635.74 USD InStock
76237-186
Rat SCF ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat SCF AccuSignal ELISA Kit
  • Natural and recombinant rat SCF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: Q26-A190
Stem Cell Factor (also known as SCF, kit-ligand, KL, or steel factor) is a cytokine that binds to the c-Kit receptor (CD117). It is mapped to 12q21.32. SCF was primarily expressed by perivascular cells throughout the bone marrow, and it plays a role in the regulation of HSCs in the stem cell niche in the bone marrow. SCF can exist both as a transmembrane protein and a soluble protein. This gene plays an important role in the hematopoiesis during embryonic development. During development, the presence of the SCF also plays an important role in the localization of melanocytes, cells that produce melanin and control pigmentation. In addition to it, SCF can promote mast cell adhesion, migration, proliferation, and survival. It also promotes the release of histamine and tryptase, which are involved in the allergic response.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml rat SCF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma (EDTA) to each empty well. It is recommended that each rat SCF standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty