Would you like to log in again?
Mouse TPO ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0310
76236-782EA 635.74 USD InStock
76236-782
Mouse TPO ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse TPO AccuSignal ELISA Kit
  • Natural and recombinant mouse TPO.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S22-T356
Thrombopoietin(TPO) is also called megakaryocyte growth and development factor(MGDF). TPO, a lineage-specific cytokine affecting the proliferation and maturation of megakaryocytes from committed progenitor cells, is believed to be the major physiological regulator of circulating platelet levels. Human, dog, and mouse cDNAs for MGDF are highly conserved and encode open reading frames for proteins of 353, 352, and 356 amino acids, respectively, including predicted signal peptides. MGDF is a novel cytokine that regulates megakaryocyte development and is a ligand for the MPL receptor. The human MGDF gene has been mapped to chromosome 3q26.3. The standard product used in this kit is gene recombinant mouse TPO, consisting of 419 amino acids with the molecular mass of 35KDa. As a result of glycosylation, the molecular mass is 75KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse TPO standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse ell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse TPO standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty