Would you like to log in again?
Mouse CCL24 - Eotaxin-2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0352
76237-610EA 635.74 USD InStock
76237-610
Mouse CCL24 - Eotaxin-2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CCL24 - Eotaxin-2 AccuSignal ELISA Kit
  • Natural and recombinant mouse CCL24.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: V27-V119
Chemokine(C-C motif) ligand 24(CCL24) also known as myeloid progenitor inhibitory factor 2(MPIF-2) or eosinophil chemotactic protein 2(eotaxin-2) is a protein that in humans is encoded by the CCL24 gene.1 By use of PCR analysis of somatic cell hybrid DNAs, radiation hybrid mapping, and a chromosome 7-specific YAC library, mapped the SCYA24 gene to chromosome 7q11.23.2 CCL24 is a small cytokine belonging to the CC chemokine family. CCL24 interacts with chemokine receptor CCR3 to induce chemotaxis in eosinophils.3 This chemokine is also strongly chemotactic for resting T lymphocytes and slightly chemotactic for neutrophils.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse CCL24 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CCL24 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty