Would you like to log in again?
Mouse Gp130 - IL6ST ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0353
76236-852EA 635.74 USD InStock
76236-852
Mouse Gp130 - IL6ST ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Gp130 - IL6ST AccuSignal ELISA Kit
  • Natural and recombinant mouse Gp130.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q23-E617
GP130(Glycoprotein 130) also known as IL6ST or CD130, is a transmembrane protein which is the founding member of the class of all cytokine receptors. The GP130 gene is mapped on 5q11.2. Gp130 showed an apparent molecular mass of 130 kD by SDS-PAGE. Gp130 is involved in the formation of high-affinity IL6-binding sites and in IL6 signal transduction. Sgp130 has a role in modulating signals transduced by membrane-bound gp130. Gp130 is an important part of many different types of signaling complexes.Gp130 molecules with mutations in either of these segments could not transduce a growth signal.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 8000pg/ml, 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml mouse Gp130 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each mouse Gp130 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty