Would you like to log in again?
Human Growth Hormone ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0354
76236-854EA 723.6 USD InStock
76236-854
Human Growth Hormone ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Growth Hormone AccuSignal ELISA Kit
  • Natural and recombinant human GH.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: F27-F217
Growth hormone(GH) is synthesized by acidophilic or somatotropic cells of the anterior pituitary gland. Human growth hormone has a molecular mass of 22,005 and contains 191 amino acid residues with 2 disulfide bridges. The human genes for growth hormone(GH), has been located on chromosome 17 in humans. GH replacement of adults with acquired GH deficiency(GHD) results in body composition changes including increases in lean mass and bone mineral density. However, the effects of long-term GH therapy on cognitive function are largely unknown, and there are conflicting data regarding quality of life. The standard product used in this kit is recombinant human Growth Hormone, consisting of 192 amino acids with the molecular mass of 22KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml human GH standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human GH standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty