Would you like to log in again?
Human Resistin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0355
76236-856EA 723.6 USD InStock
76236-856
Human Resistin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Resistin AccuSignal ELISA Kit
  • Natural and recombinant human Resistin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: K19-P108
Resistin is an adipose-derived hormone postulated to link adiposity to insulin resistance.1 Type 2 diabetes, characterized by target-tissue resistance to insulin, is epidemic in industrialized societies and is strongly associated with obesity. Adipocytes secrete a unique signalling molecule, which called resistin(for resistance to insulin), that may be the hormone potentially links obesity to diabetes.2 Resistin is overexpressed in human adipose tissue of obese individuals and is likely to modulate insulin sensitivity. Resistin is, therefore, a candidate gene for insulin resistance.3 The standard product used in this kit is human Resistin with the molecular mass of 51-53KDa. The detected Resistin includes zymogen and active enzyme.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml, 78pg/ml human Resistin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin , EDTA) to each empty well. We recommend that each human Resistin standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty