Would you like to log in again?
Mouse VEGFR2 - KDR ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0360
76236-866EA 686.86 USD InStock
76236-866
Mouse VEGFR2 - KDR ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse VEGFR2 - KDR AccuSignal ELISA Kit
  • Natural and recombinant mouse VEGFR2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A20-E762
Vascular endothelial growth factor receptor 2(VEGFR-2) is a VEGF receptor. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. It is mapped to 4q12. This gene encodes one of the two receptors of the VEGF. It functions as the main mediator of VEGF-induced endothelial proliferation, survival, migration, tubular morphogenesis and sprouting. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc. VEGF receptor 2 and the adherens junction act as shear-stress cotransducers, mediating the transduction of shear-stress signals into vascular endothelial cells. In addition to those, VEGFR-2 is also a positive functional marker defining stem cells and distinguishing them from progenitors.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse VEGFR2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates or serum to each empty well. It is recommended that each mouse VEGFR2 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty