Would you like to log in again?
Human Survivin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0369
76237-204EA 724 USD InStock
76237-204
Human Survivin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Survivin AccuSignal ELISA Kit
  • Natural and recombinant human Survivin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: M1-D142
Survivin(also termed API4) belongs to the family of genes known as inhibitors of apoptosis, and it has been implicated in both prevention of cell death and control of mitosis. Survivin is prominently expressed in transformed cell lines and in all the most common human cancers of lung, colon, pancreas, prostate and breast in vivo. It is also found in approximately 50% of high-grade non-Hodgkin's lymphomas. Survivin encodes a deduced 142-amino acid protein and localizes on chromosome 17q25. The overexpression of it in cancer may overcome an apoptotic checkpoint and favour aberrant progression of transformed cells through mitosis. The standard product used in this kit is recombinant human Survivin with the molecular mass of 34 KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human Survivin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Survivin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty