Would you like to log in again?
Human APP ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0375
76237-216EA 687 USD InStock
76237-216
Human APP ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human APP AccuSignal ELISA Kit
  • Natural and recombinant human APP.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L18-L688
Amyloid precursor protein(APP) is an integral membrane protein expressed in many tissues and concentrated in the synapses of neurons. Its primary function is not known, though it has been implicated as a regulator of synapse formation, neural plasticity and iron export. APP is best known and most commonly studied as the precursor molecule whose proteolysis generates beta amyloid(Abeta), a 39- to 42-amino acid peptide whose amyloid fibrillar form is the primary component of amyloid plaques found in the brains of Alzheimer's disease patients. APP undergoes posttranslational proteolytic processing by alpha-, beta-, and gamma-secretases. Alpha-secretase generates soluble amyloid protein, while beta- and gamma-secretases generate APP components with amyloidogenic features. These 2 processing pathways are mutually exclusive.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human APP standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human APP standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty