Would you like to log in again?
Human BAFF ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0380
76237-226EA 686.86 USD InStock
76237-226
Human BAFF ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human BAFF AccuSignal ELISA Kit
  • Natural and recombinant human BAFF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A134-L285
BAFF was regularly detected by enzyme-linked immunosorbent assay in brain tissue lysates and in normal spinal fluid, and in astrocytes by double fluorescence microscopy. BAFF was localized in astrocytes close to BAFF-R-expressing immune cells. BAFF receptors were strongly expressed in situ in primary central nervous system(CNS) lymphomas.1 The TNF superfamily member B cell-activating factor(BAFF) plays an important role in humoral immunity and in autoimmune diseases, including RA.Local BAFF gene targeting inhibited proinflammatory cytokine expression, suppressed generation of plasma cells and Th17 cells, and markedly ameliorated joint pathology.2 The B cell activating factor BAFF(BlyS/TALL-1/zTNF4) is a tumor necrosis factor(TNF)-related ligand that promotes B cell survival and binds to three receptors(BCMA, TACI, and the recently described BAFF-R).3 Human BAFF was mapped to chromosome 13q32-34.4 The standard used in this kit is recombinant soluble human BAFF(A134-L295) with the molecular mass of 19.6KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human BAFF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human BAFF standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty