Would you like to log in again?
Human P-Cadherin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0382
76237-230EA 686.86 USD InStock
76237-230
Human P-Cadherin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human P-Cadherin AccuSignal ELISA Kit
  • Natural and recombinant human P-Cadherin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: D108-G654
Cadherins are calcium-dependent cell-cell adhesion molecules that mediate cell-cell binding in a homophilic manner. They play an important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. Cadherin expression is regulated spatially as well as temporally. Cadherins are thought to play an important role in development and maintenance of tissues through selective cell-cell adhesion activity and may be involved also in the invasion and metastasis of malignant tumors. Cadherin regulates dendritic spine morphogenesis. A cadherin gene cluster is mapped to a region of chromosome 5 subject to frequent allelic loss in carcinoma. The standard product used in this kit is recombinant P-Cadherin with the molecular mass of 120-130Kda after glycosylation.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human P-Cadherin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. It is recommended that each human P-Cadherin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty