Would you like to log in again?
Human Cathepsin L ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0389
76237-244EA 686.86 USD InStock
76237-244
Human Cathepsin L ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Cathepsin L AccuSignal ELISA Kit
  • Natural and recombinant human Cathepsin L.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E113-V333
Cathepsin L is an important lysosomal endopeptidase enzyme which is involved in the initiation of protein degradation. It also shows the most potent collagenolytic and elastinolytic activity in vitro of any of the cathepsins. The gene is mapped to 9q21-q22. It is a member of the Peptidase C1 family, which play an important role in diverse processes including normal lysosome mediated protein turnover, antigen and proprotein processing, and apoptosis. Cathepsin L has been reported in many organisms including fish, birds and mammals.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human Cathepsin L standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Cathepsin L standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty