Would you like to log in again?
Human ESM1 - Endocan ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0444
76235-606EA 636 USD InStock
76235-606
Human ESM1 - Endocan ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human ESM1 - Endocan AccuSignal ELISA Kit
  • Natural and recombinant human ESM1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: W20-R184
Endothelial cell-specific molecule 1, also known as Endocan, is a protein that in humans is encoded by the ESM1 gene. This gene encodes a secreted protein which is mainly expressed in the endothelial cells in human lung and kidney tissues. The expression of this gene is regulated by cytokines, suggesting that it may play a role in endothelium-dependent pathological disorders. The transcript contains multiple polyadenylation and mRNA instability signals. ESM1 has been described as a specific biomarker of tip cells during neoangiogenesis by independent teams. Its expression has been shown to be increase in presence of pro-angiogenic growth factors such as VEGF(vascular endothelial growth factor) or FGF-2 (fibroblast growth factor 2).
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human ESM1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human ESM1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty