Would you like to log in again?
Mouse IL-1RA - IL1RN ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0462
76237-310EA 642 USD InStock
76237-310
Mouse IL-1RA - IL1RN ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Interleukin-1RA - IL1RN AccuSignal ELISA Kit
  • Natural and recombinant mouse IL-1RA.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.colI; Immunogen sequence: R27-Q178
The interleukin-1 receptor antagonist(IL-1RA) is a protein that in humans is encoded by the IL1RN gene. The human IL1RN gene was localized to the long arm of chromosome 2 at band 2q14.2 by fluorescence in situ hybridization. IL-1RA is a member of the interleukin 1 cytokine family. This protein inhibits the activities of interleukin 1, alpha(IL1A) and interleukin 1, beta(IL1B), and modulates a variety of interleukin 1 related immune and inflammatory responses.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse IL-1RA standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IL-1RA standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty