Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Mouse IL-17C ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0466
76237-318EA 845.49 USD InStock
76237-318
Mouse IL-17C ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Interleukin-17C AccuSignal ELISA Kit
  • Natural and recombinant mouse IL-17C.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.colI; Immunogen sequence: D17-Q194
IL17C, also known as CX2, is a protein that in humans is encoded by the IL17C gene. IL17C is mapped to 16q24.3. The protein encoded by this gene is a T cell-derived cytokine that shares the sequence similarity with IL17. This cytokine was reported to stimulate the release of tumor necrosis factor alpha and interleukin 1 beta from a monocytic cell line. The expression of this cytokine was found to be restricted to activated T cells. IL17C is an essential autocrine cytokine regulating innate epithelial immune responses. It also plays an important role in the pathogenesis of inflammatory arthritis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 8000pg/ml, 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml mouse IL-17C standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IL-17C standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty