Would you like to log in again?
Human TFPI ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0504
76237-368EA 635.74 USD InStock
76237-368
Human TFPI ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human TFPI AccuSignal ELISA Kit
  • Natural and recombinant human TFPI.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: D29-K282
TFPI(Tissue factor pathway inhibitor),also known as EPI, LACI or TFPI1, is a single-chain polypeptide which can reversibly inhibit Factor Xa(Xa) and Thrombin(Factor IIa). The gene for TFPI is located on chromosome 2q31-q32.1, and has nine exons which span 70 kb. TFPI consists of a highly negatively-charged amino-terminus, three tandemly-linked Kunitz domains, and a highly positively-charged carboxy-terminus.TFPI contributes significantly to the inhibition of Xa in vivo, despite being present at concentrations of only 2.5 nM.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human TFPI standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA) or urine to each empty well. It is recommended that each human TFPI standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty