Would you like to log in again?
Human CD10 - Neprilysin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0512
76235-644EA 636 USD InStock
76235-644
Human CD10 - Neprilysin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CD10 - Neprilysin AccuSignal ELISA Kit
  • Natural and recombinant human CD10.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: Y52-W750
CD10, also known as membrane metallo-endopeptidase, neutral endopeptidase(NEP), Neprilysin, or common acute lymphoblastic leukemia antigen(CALLA), is a zinc-dependent metalloprotease enzyme that degrades a number of small secreted peptides, most notably theamyloid beta peptide whose abnormal misfolding and aggregation in neural tissue has been implicated as a cause of Alzheimer's disease. This gene is localized to human chromosome 3 by study of somatic cell hybrids and regionalized the location to 3q21-q27 by in situ hybridization. By cDNA transfection analysis, CD10 is confirmed as a functional neutral endopeptidase of the type that has previously been called enkephalinase. CD10 has also been called atriopeptidase. Atriopeptidase specifically degrades atrial natriuretic factor. A specific enzyme inhibitor was developed and reported that it had effects similar to those of low-dose ANF infusion. These effects include diuresis, natriuresis, vasodilatation, and suppression of the renin-angiotensin-aldosterone system.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 8000pg/ml, 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml human CD10 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CD10 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty