Would you like to log in again?
Human Entactin - NID-1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0517
76236-922EA 635.74 USD InStock
76236-922
Human Entactin - NID-1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Entactin - NID-1 AccuSignal ELISA Kit
  • Natural and recombinant human Nidogen-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L29-K1114
Nidogen-1(NID-1), also known as Entactin, is a protein that in humans is encoded by the NID1 gene. It is a member of the nidogen family of basement membrane glycoproteins. This gene is mapped to 1q42.3. Nidogen-1 is a component of the basement membrane alongside other components such as collagen type IV, proteoglycans(heparan sulfate and glycosaminoglycans), laminin and fibronectin. The protein interacts with several other components of basement membranes. Structurally it(along with perlecan) connects the networks formed by collagens and laminins to each other. It may also play a role in cell interactions with the extracellular matrix. Nidogen-1 also can serve as a bridge between the 2 most abundant molecules in the basement membrane: type IV collagen and laminin.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human Nidogen-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. It is recommended that each human Nidogen-1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty