Would you like to log in again?
Human PEDF - SerpinF1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0537
76235-670EA 635.74 USD InStock
76235-670
Human PEDF - SerpinF1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human PEDF - SerpinF1 AccuSignal ELISA Kit
  • Natural and recombinant human PEDF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: M1-P418
Pigment epithelium-derived factor (PEDF) is also known as serpin F1 (SERPINF1). In humans, it is encoded by the SERPINF1 gene. PEDF has a variety of functions including antiangiogenic, antitumorigenic, and neurotrophic properties. It suppresses retinal neovascularization and endothelial cell proliferation. And Antiangiogenic function is also conferred by PEDF through inhibition of both VEGFR-1 and VEGFR-2. In addition, the antitumorigenic effects of PEDF are not only due to inhibition of supporting vasculature, but also due to effects on the cancer cells themselves. PEDF is shown to inhibit cancer cell proliferation and increase apoptosis via the FAS/FASL pathway. Expression of PEDF in the human retina is found at 7.4 weeks of gestation, suggesting it may play a role in retinal neuron differentiation.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml human PEDF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human PEDF standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty