Would you like to log in again?
Human CEA ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0544
76237-384EA 723.6 USD InStock
76237-384
Human CEA ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CEA AccuSignal ELISA Kit
  • Natural and recombinant human CEA.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: K35-A685
Carcinoembryonic antigen(CEA) is a complex immunoreactive glycoprotein with a molecular weight of 180,000 comprising 60% carbohydrate. It is found in adenocarcinomas of endodermally derived digestive system epithelia and in fetal colon.Carcinoembryonic antigen is one of the most widely used tumor markers in serum immunoassay determinations of carcinoma.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human CEA standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma( heparin, EDTA) to each empty well. It is recommended that each human CEA standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty